Learn More
ACROBiosystems FITC-Labeled Human CD7 Protein, Llama IgG2b Fc Tag
Catalog No. 502019567
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human CD7, Llama IgG2b Fc Tag (CD7-HF257) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). It is the FITC labeled form of Human CD7, Llama IgG2b Fc Tag, low endotoxin (CD7-H5258).;This protein carries a llama IgG2b Fc tag at the N-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9567 Supplier ACROBiosystems Supplier No. CD7HF25725UG
Recombinant Protein;25UG;FITC-Labeled Human CD7, Llama IgG2b Fc Tag (CD7-HF257) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). It is the FITC labeled form of Human CD7, Llama IgG2b Fc Tag, low endotoxin (CD7-H5258).;This protein carries a llama IgG2b Fc tag at the N-terminus. The protein has a calculated MW of 44.3 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.