Learn More
ACROBiosystems FITC-Labeled Human CD7 Protein, His Tag
Catalog No. 502019569
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human CD7, His Tag (CD7-HF2H6) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). It is the FITC labeled form of Human CD7, His Tag (CD7-H52H7).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.3 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-201-9569 Supplier ACROBiosystems Supplier No. CD7HF2H625UG
Recombinant Protein;25UG;FITC-Labeled Human CD7, His Tag (CD7-HF2H6) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). It is the FITC labeled form of Human CD7, His Tag (CD7-H52H7).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.3 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.