Learn More
Creative Biomart Recombinant Protein 28.8 kDa Host-E.coli Inquiry 50 UG 1/EA
Catalog No. 5000555553
Encompass
mCherry is the second generation monomeric red fluorescent protein that have improved brightness and photostability The recombinant mCherry is expressed and purified from transformed E coli using a method that ensures high purity and maximal fluorescence intensity The protein is a 28 8 kDa monomer with 256 amino acids pI 6 23 Ex 587 nm (540-590 nm) Em 610 nm (550-650 nm) The protein is engineered with 6xHis-tag on the N-terminus
Catalog No. 50-005-55553 Supplier Creative Biomart Supplier No. MCHERRY5
mCherry is the second generation monomeric red fluorescent protein that have improved brightness and photostability The recombinant mCherry is expressed and purified from transformed E coli using a method that ensures high purity and maximal fluorescence intensity The protein is a 28 8 kDa monomer with 256 amino acids pI 6 23 Ex 587 nm (540-590 nm) Em 610 nm (550-650 nm) The protein is engineered with 6xHis-tag on the N-terminus