Learn More
ACROBiosystems Human MSP1D1 Protein, His Tag
Supplier:  ACROBiosystems APOH51H330UG
Encompass_Preferred
Recombinant Protein;30UG;Human MSP1D1, His Tag (APO-H51H3) is expressed from E.coli cells. It contains AA Gly 1- Gln 211.;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 24.7 kDa. The protein migrates as 24 kDa under reducing (R) condition (SDS-PAGE). This protein is used together with CD20 nanodisc protein (CD0-H52H1) as isotype control.;2e5 of CD20-CAR-293 cells transfected with anti-CD20-scFv were stained with 100 μL of 10 μg/mL of Human MSP1D1, His Tag (Cat. No. APO-H51H3) and negative control antibody respectively, washed and then followed by PE anti-His antibody and analyzed with FACS (QC tested).
Catalog No. 50-201-9816