Learn More
ACROBiosystems Cynomolgus CD20 / MS4A1 Full Length Protein, His Tag
Supplier:  ACROBiosystems CD0C52H8500UG
Encompass_Preferred
Recombinant Protein;500UG;Cynomolgus CD20 Full Length, His Tag (CD0-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Pro 297 (Accession # M4ZHZ6-1 ).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.2 kDa. The protein migrates as 36-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Rituximab at 2 μg/mL (100 μL/well) can bind Cynomolgus CD20 Full Length, His Tag (Cat. No. CD0-C52H8) with a linear range of 1-31 ng/mL (QC tested).;Measured by its binding ability in a functional ELISA. Immobilized Ofatumumab at 2 μg/mL (100 μL/well) can bind Cynomolgus CD20 Full Length, His Tag (Cat. No. CD0-C52H8) with a linear range of 0.2-16 ng/mL (QC tested).
Catalog No. 50-201-9454
Add to Cart
Request a Quote