Learn More
ACROBiosystems Human Carbonic Anhydrase IX / CA9 (38-414) Protein, His Tag
Supplier: ACROBiosystems CA9H522650UG
Recombinant Protein;50UG;Human Carbonic Anhydrase IX (38-414), His Tag (CA9-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - Asp 414 (Accession # Q16790-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.