Learn More
ACROBiosystems Human CD8 alpha / CD8A Protein, His Tag
Supplier:  ACROBiosystems CDAH52H350UG
Encompass_Preferred
Recombinant Protein;50UG;Human CD8 alpha, His Tag (CDA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (Accession # P01732-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.5 kDa. The protein migrates as 22-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD8 alpha, His Tag (Cat. No. CDA-H52H3) at 1 μg/mL (100 μL/well) can bind Anti-CD8 alpha Antibody, Mouse IgG2a (clone: OKT8) with a linear range of 0.2-39 ng/mL (QC tested).;Immobilized Anti-CD8 alpha Antibody, Mouse IgG2a (clone: OKT8) at 1 μg/mL (100 μL/well) can bind Human CD8 alpha, His Tag (Cat. No. CDA-H52H3) with a linear range of 0.6-10 ng/mL (Routinely tested).
Catalog No. 50-201-9726