Learn More
ACROBiosystems Mouse IL-33 Protein, His Tag
Supplier:  ACROBiosystems IL3M524550UG
Encompass_Preferred
Recombinant Protein;50UG;Mouse IL-33, His Tag (IL3-M5245) is expressed from human 293 cells (HEK293). It contains AA Ser 109 - Ile 266 (Accession # Q8BVZ5-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 19.5 kDa.
Catalog No. 50-201-9483