Learn More
Sino Biological Recombinant Protein A
Supplier:  Sino Biological 10600P07E10
Encompass_Preferred
The properties of protein A enables it as a powerful affinity ligand for several immunological and purification applications. High selectivity and good physiochemical stability have made protein A the preferred generic ligand for affinity purification of antibodies and molecules tagged with an antibody Fc-region. Protein A can also be used in various immunochemical assays including Western blotting, immunohistochemistry, and ELISA applications by conjugation with different reporter molecules, such as fluorescent dyes (FITC), enzyme markers (peroxidase, ?-galactosidase, alkaline phosphatase), biotin, and colloidal gold. Immunoprecipitation studies with protein A conjugated to beads are also commonly used to purify proteins or protein complexes indirectly through antibodies against the protein or protein complex of interest.
Catalog No. 50-237-0859