Learn More
G-Biosciences Recombinant Protein A/G

Description
- Recombinant Protein A/G binds well to IgG subclasses but does not bind IgA, IgM or serum albumin.
- This makes Protein A/G an excellent tool for purification and detection of monoclonal antibodies from IgG subclasses, without interference from IgA, IgM and serum albumin.
- Individual subclasses of monoclonals are likely to have a stronger affinity to the chimeric Protein A/G than to either Protein A or Protein G.
Specifications
Specifications
| Product Line | HiTrap |
| Column Type | Affinity |
| Format | Bottle |
| Stationary Phase | Protein A/G |
| Sterility | Non-sterile |
| Volume (Metric) | 5 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.