Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Recombinant Protein A/G
SCI_ED

Catalog No. S03101
Change view
Click to view available options
Volume (Metric):
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S03101 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03101 Supplier G-Biosciences Supplier No. 7862012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein A/G contains four Fc-binding domains from Protein A and two from Protein G but lacks the albumin binding sites.

  • Recombinant Protein A/G binds well to IgG subclasses but does not bind IgA, IgM or serum albumin.
  • This makes Protein A/G an excellent tool for purification and detection of monoclonal antibodies from IgG subclasses, without interference from IgA, IgM and serum albumin.
  • Individual subclasses of monoclonals are likely to have a stronger affinity to the chimeric Protein A/G than to either Protein A or Protein G.

Specifications

Product Line HiTrap
Column Type Affinity
Format Bottle
Stationary Phase Protein A/G
Sterility Non-sterile
Volume (Metric) 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.