Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novex™ Recombinant Protein A-Sepharose™ 4B

Catalog No. 101142
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-114-2 25 mL
10-114-1 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-114-2 Supplier Novex™ Supplier No. 101142
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein A-Sepharose™ 4B

This product is prepared by covalently coupling highly purified Staph. aureus Cowan strain Protein A produced in E. Coli to Sepharose™ 4B.

This product is supplied as a 50% suspension consisting of recombinant Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% ).

Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A Sepharose
Column Type Affinity
Stationary Phase Protein A
Content And Storage 2°C to 8°C. Do Not Freeze.
This product is supplied as a 50% suspension (25 ml Recombinant Protein A-Sepharose™ 4B + 25 ml PBS) Wet Ice
Quantity 25 mL
Form Suspension
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.