Learn More
Abcam Recombinant Protein G, 10MG
Supplier:  Abcam AB15572410MG
Encompass_Preferred
Recombinant Protein G is a Streptococcus sp. 'group G' Fragment protein, expressed in Escherichia coli, with >98% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: Immunoglobulin G-binding protein G, IgG-binding protein G, spg
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1929172