Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Recombinant Protein G - Sepharose™ 4B

Catalog No. 101242
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-124-2 5 mL
10-124-1 2 mL
10-124-3 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 10-124-2 Supplier Invitrogen™ Supplier No. 101242
  • $736.00
    $684.65 / Each of 1

    Save $51.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein G - Sepharose™ 4B

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This product is prepared by covalently coupling highly purified rec-Protein G to Sepharose™ 4B. It is supplied as a 50% suspension containing rec-Protein G-Sepharose™ 4B + PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin affinity purification and Immunoprecipitation.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Product Type Protein G Sepharose
    Product Line Sepharose
    Column Type Affinity
    Stationary Phase Protein G
    Content And Storage This product is supplied as a 50% suspension (5 ml Recombinant Protein G-Sepharose™ 4B + 5 ml PBS) Wet Ice
    Quantity 5 mL
    Form Suspension
    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.