Learn More
Invitrogen™ Recombinant Protein G - Sepharose™ 4B
Description
Applications: Immunoglobulin affinity purification and Immunoprecipitation.
Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:
Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
Specifications
Specifications
| Product Type | Protein G Sepharose |
| Product Line | Sepharose |
| Column Type | Affinity |
| Stationary Phase | Protein G |
| Content And Storage | This product is supplied as a 50% suspension (25 ml Recombinant Protein G-Sepharose™ 4B + 25 ml PBS) Wet Ice |
| Quantity | 25 mL |
| Form | Suspension |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.