Learn More
Creative Biomart Recombinant Protein Inquiry Host-Blood cell Species - Human 50 UG 1/EA
Catalog No. 5000555508
Encompass
Purified from human blood cells by fractination The highest activity soluble and homogenous Consists of a high purity mixture of 26S proteasomes singly (26S) and doubly (30S) capped with 19S regulatory subunit complexes in the ratio of 40 % single cap 60 % double capped at the time of preparation
Catalog No. 50-005-55508 Supplier Creative Biomart Supplier No. PROTEASOME 26S38H
Purified from human blood cells by fractination The highest activity soluble and homogenous Consists of a high purity mixture of 26S proteasomes singly (26S) and doubly (30S) capped with 19S regulatory subunit complexes in the ratio of 40 % single cap 60 % double capped at the time of preparation