Learn More
Creative Biomart Recombinant Protein Inquiry Host-Plasma Species - Human 1 ML 1/EA
Catalog No. 5000555451
Encompass
This preparation of IgE ( ) is provided as neat human plasma in EDTA prepared by the addition of purified human IgE ( ) IgE activity is 15 000 IU/ml IgE content is reported upon assay Normal Human Plasma spiked with Cell Culture Human IgE lambda
Catalog No. 50-005-55451 Supplier Creative Biomart Supplier No. IGE18H
This preparation of IgE ( ) is provided as neat human plasma in EDTA prepared by the addition of purified human IgE ( ) IgE activity is 15 000 IU/ml IgE content is reported upon assay Normal Human Plasma spiked with Cell Culture Human IgE lambda