Learn More
Sino Biological Recombinant Protein L
Supplier:  Sino Biological 11044H07E1
Encompass_Preferred
Protein L was first isolated from the surface of bacterial species Peptostreptoccus magnus and was found to bind Ig through L chain interaction, from which the name was suggested.
Catalog No. 50-237-3008