Learn More
Abcam Recombinant Rabies Virus Glycoprotein (His tag), 100UG
Supplier:  Abcam AB225663100UG
Encompass_Preferred
Recombinant Rabies Virus Glycoprotein (His tag) is a Rabies virus (strain ERA) Fragment protein, in the 20 to 459 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE. The predicted molecular weight of ab225663 protein is 65.6 kDa. - Save time and ensure accurate results - use our recombinant Rabies Virus Glycoprotein (RABV G) as a control - Contains N-terminal 6x His-SUMO-tag
Protein/ Peptide/ Enzyme Synonyms: Glycoprotein, G
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2040419