Learn More
Abcam Recombinant rat Cyclophilin F protein (Active), 20UG
Catalog No. 502676534
Encompass_Preferred
Recombinant rat Cyclophilin F protein (Active) is a Rat Full Length protein, in the 29 to 206 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, FuncS, MS.
Protein/ Peptide/ Enzyme Synonyms: PPIase F, Cyclophilin D, Cyclophilin F, Rotamase F, CyP-D, CypD, Ppif
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6534 Supplier Abcam Supplier No. AB21942220UG
Recombinant rat Cyclophilin F protein (Active) is a Rat Full Length protein, in the 29 to 206 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, FuncS, MS.
Protein/ Peptide/ Enzyme Synonyms: PPIase F, Cyclophilin D, Cyclophilin F, Rotamase F, CyP-D, CypD, Ppif
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement