Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&DSystems™ Recombinant Rat EGFR mFc Chimera Protein, CF

Catalog No. 11522ER050
Change view
Click to view available options
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
11-522-ER050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-522-ER050 Supplier R&DSystems™ Supplier No. 11522ER050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Recombinant Rat EGFR mFc Chimera binds to Recombinant Rat EGF Protein (Catalog # 3214-EG ) with an ED50 of 0.250-2.50 μg/mL.

Specifications

Accession Number NP_113695.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 1956
Molecular Weight (g/mol) M.W.-observed: 110-130 kDa, under reducing conditions., M.W.-theroretical: 97 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat EGFR protein Rat EGFR (Leu25-Ile647) (N-terminus) Accession # NP_113695.1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EGFR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.