Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat GDNF Protein

Catalog No. 512GF01M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-2GF-01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-2GF-01M Supplier R&D Systems™ Supplier No. 512GF01M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity

Measured in a cell proliferation assay using SH-SY5Y human neuroblastoma cells. The ED50 for this effect is 2-12 ng/mL in the presence of Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ). Measured by its binding ability in a functional ELISA. Immobilized Recombinant Rat GFR alpha-1/GDNF R alpha-1 Fc Chimera (Catalog # 560-GR ) at 1 μg/mL binds Recombinant Rat GDNF with an apparent Kd <4 nM.

Specifications

Accession Number Q07731
Characterization Bioactivity, Binding Activity
For Use With (Application) Bioactivity, Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 2668
Molecular Weight (g/mol) M.W. (theoretical): 15 kDa (monomer)
Quantity 1 mg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived rat GDNF protein Ser78-Ile211
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Alias Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Gene Symbol Gdnf
Product Type Recombinant Protein
Conjugate Unconjugated
Species Rat
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein GDNF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.