Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat GITR/TNFRSF18 Fc Chimera Protein

Catalog No. 9130GR050
Change view
Click to view available options
:
Quantity: 50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
91-30G-R050 Quantity: 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-30G-R050 Supplier R&D Systems™ Supplier No. 9130GR050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Rat GITR/TNFRSF18 Fc Chimera is immobilized at 2 μg/mL, 100 μL/well, the concentration of Recombinant Mouse GITR Ligand/TNFSF18 aa 47-173 (Catalog # 2177-GL ) that produces 50% of the optimal binding response is 25-150 ng/mL

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number NP_001019520
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 8784
Molecular Weight (g/mol) M.W. (Observed): 55-64 kDa, reducing conditions; M.W. (theoretical): 41 kDa
Name FGF10, FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF-2, produced by fibroblasts of urinary bladder lamina propria
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat GITR/TNFRSF18 protein Rat GITR/TNFRSF18 (Glu25-His153) (N-terminus) Accession # NP_001019520 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Biological Activity 25 to 150ng/mL (ED 50)
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag No
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GITR/TNFRSF18
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.