Learn More
Abcam Recombinant rat IL-1 beta protein (Active), 50UG
Catalog No. 502655506
Encompass_Preferred
Recombinant rat IL-1 beta protein (Active) is a Rat Fragment protein, in the 117 to 268 aa range, expressed in HEK 293, with >95% purity, <= 0.005 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS, MS, HPLC.
Protein/ Peptide/ Enzyme Synonyms: Interleukin-1 beta, IL-1 beta, Il1b
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-5506 Supplier Abcam Supplier No. AB28180750UG
Recombinant rat IL-1 beta protein (Active) is a Rat Fragment protein, in the 117 to 268 aa range, expressed in HEK 293, with >95% purity, <= 0.005 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS, MS, HPLC.
Protein/ Peptide/ Enzyme Synonyms: Interleukin-1 beta, IL-1 beta, Il1b
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement