Learn More
R&D Systems™ Recombinant Rat IL-17A/F Heterodimer Protein
Description
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Accession Number | Q61453 (IL-17A), Q5BJ95 (IL-17F) |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein |
| Molecular Weight (g/mol) | 15 kDa |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived rat IL-17A/F Heterodimer protein, (N-terminus)Rat IL-17A (Ala18-Ser150 (Ile46Leu)) Accession &Num; Q61453 Rat IL-17F (Arg29-Ala161) Accession &Num; Q5BJ95 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Structural Form | Disulfide-linked heterodimer |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method |
| Biological Activity | 3 to 18ng/mL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.