Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat NKp30 Fc Chimera Protein

Catalog No. 10057NK050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10057NK050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10057NK050 Supplier R&D Systems™ Supplier No. 10057NK050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its binding ability in a functional ELISA. When Recombinant Rat NKp30/NCR3 Fc Chimera is immobilized at 5 μg/mL (100 μL/well), the concentration of Biotinylated Recombinant Human B7-H6 Fc Chimera (Catalog # BT7144 ) that produces 50% of the optimal binding response is 1-6 μg/mL.

Specifications

Accession Number Q8CFD9
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MES and NaCl.
Gene ID (Entrez) 259197
Molecular Weight (g/mol) M.W. (Observed): 42-64 kDa, reducing conditions; M.W. (theoretical): 41 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived rat NKp30/NCR3 protein Rat NKp30 (Val22-Ser150) (N-terminus) Accession # Q8CFD9 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Specific Reactivity NKp30/NCR3
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in MES and NaCl.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein NKp30/NCR3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.