Learn More
R&D Systems™ Recombinant Rat Pentraxin 2/SAP Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 29339 |
| Molecular Weight (g/mol) | 25 kDa |
| Name | Pentraxin 2/SAP |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived rat Pentraxin 2/SAP protein Gln21-Ser228 with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked homopentamer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.