Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Periostin/OSF-2 Protein

Catalog No. 8994F2050
Change view
Click to view available options
50μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-94F-2050 50μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-94F-2050 Supplier R&D Systems™ Supplier No. 8994F2050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat Periostin/OSF-2 Protein is derived from HEK293. The Recombinant Rat Periostin/OSF-2 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number D3ZAF5
For Use With (Application) Bioactivity
Gene ID (Entrez) 10631
Molecular Weight (g/mol) Observed M.W.: 77-95kDa, reducing conditions, Theoretical M.W.: 87kDa
Quantity 50μg
Source Human embryonic kidney cell,HEK293-derived rat Periostin/OSF-2 protein Asn24-Ser800,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Fasciclin I-like, MGC119510, MGC119511, OSF2, OSF-2, OSF-2osteoblast specific factor 2 (fasciclin I-like), OSF2periodontal ligament-specific periostin, Osteoblast-specific factor 2, PDLPOSTN, periostin, periostin isoform thy2, periostin isoform thy4, periostin isoform thy6, periostin isoform thy8, periostin, osteoblast specific factor, PNRP11-412K4.1, POSTN, TRIF52
Common Name Periostin/OSF-2
Gene Symbol Postn
Biological Activity Measured by its ability to induce adhesion of ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.6-3.6μg/mL.
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Asn24-Ser800, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.