Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat Reg3B Protein

Catalog No. 8288RG050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8288RG050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8288RG050 Supplier R&D Systems™ Supplier No. 8288RG050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Rat Reg3B Protein is derived from E. coli. The Recombinant Rat Reg3B Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P25031
For Use With (Application) Bioactivity
Gene ID (Entrez) 18489
Molecular Weight (g/mol) Observed M.W.: 16kDa, reducing conditions, Theoretical M.W.: 17kDa
Quantity 50 μg
Source E. coli-derived rat Reg3B protein Glu27-Gly175,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias PAP1, Reg3B
Common Name Reg3B
Gene Symbol Reg3b
Biological Activity Measured in a cell proliferation assay using RT4-D6P2T rat schwannoma cells. The ED50 for this effect is typically 0.2-1.0μg/mL.
Conjugate Unconjugated
Species Rat
Recombinant Recombinant
Expression System E. coli-derived Glu27-Gly175, with an N-terminal Met
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.