Learn More
R&D Systems™ Recombinant Rat TIM-3 Fc Chimera Protein
Description
Specifications
Specifications
| Accession Number | P0C0K5.2 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 84868 |
| Molecular Weight (g/mol) | M.W.-observed: 60-75 kDa, under reducing conditions, M.W.-theoretical: 46 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived rat TIM-3 protein Rat TIM-3 (Leu22-Ala194) (N-terminus) Accession # P0C0K5.2 IEGRMD Mouse IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD366, FLJ14428, HAVcr-2, HAVCR2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, KIM-3, SPTCL, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3, TIM3 T-cell membrane protein 3, TIMD3, TIMD-3, TIMD3KIM-3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.