Learn More
Active Motif Recombinant RXR-α (RXRA) protein
SDP
Supplier:  Active Motif 81082
Encompass
Recombinant human RXR-alpha (RXRA) protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_002948.1) with an N-terminal FLAG tag. The molecular weight of the protein is 52.1 kDa.
Catalog No. NC1851441