Learn More
R&D Systems™ Recombinant S. pyogenes CRISPR-associated Protein 9
Description
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, EDTA, Glycerol and TCEP. |
| Gene ID (Entrez) | 901176 |
| Molecular Weight (g/mol) | M.W. (Observed): 133 kDa, reducing conditions; M.W. (theoretical): 164 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived s. pyogenes CRISPR-Cas9 protein APKKKRKVGIHGVPAA S. pyogenes CRISPR-Cas9 (Asp2-Asp1368) (N-terminus) Accession # Q99ZW2 KRPAATKKAGQAKK-KKGYGRKKRRQRRRG HHHHHH (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.