Learn More
Biomatik Corporation Recombinant SaccharoMyces cerevisiae Site-specific recombinase Flp (FLP1)
Supplier:  Biomatik Corporation RPC22759
Encompass_Preferred
Recombinant SaccharoMyces cerevisiae Site-specific recombinase Flp (FLP1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: SaccharoMyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast). Target Name: FLP1. Target Synonyms: Protein Able. Accession Number: P03870. Expression Region: 1~423aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 54.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5728