Learn More
Abcam Recombinant Salmonella typhi uspA protein (His tag), 10UG
Catalog No. 502704126
Encompass_Preferred
Recombinant Salmonella typhi uspA protein (His tag) is a Salmonella typhi Full Length protein, in the 2 to 144 aa range, expressed in Mammalian, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: STY4212, t3925, uspA, Universal stress protein A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4126 Supplier Abcam Supplier No. AB26792310UG
Recombinant Salmonella typhi uspA protein (His tag) is a Salmonella typhi Full Length protein, in the 2 to 144 aa range, expressed in Mammalian, with >90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: STY4212, t3925, uspA, Universal stress protein A
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement