Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant SARS-CoV-2 BA.2.86 Spike GCN4-IZ His Protein, CF

Catalog No. 11478CV100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-478-CV100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-478-CV100 Supplier R&D Systems™ Supplier No. 11478CV100
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN ).

Specifications

Accession Number YP_009724390.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 43740568
Molecular Weight (g/mol) MolecularWeight-observed: 150-170 kDa, under reducing conditions., MolecularWeight-theroretical: 138 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike protein SARS-CoV-2 BA.2.86 Spike (Val16-Lys1211) (ins16MPLF, Thr19Ile, Arg21Thr, Leu24del, Pro25del, Pro26del, Ala27Ser, Ser50Leu, His69del, Val70del, Val127Phe, Gly142Asp, Tyr144del, Phe157Ser, Arg158Gly, Asn211del, Leu212Ile, Val213Gly, Leu216Phe, His245Asn, Ala264Asp, Ile332Val, Gly339His, Lys356Thr, Ser371Phe, Ser373Pro, Ser375Phe, Thr376Ala, Arg403Lys, Asp405Asn, Arg408Ser, Lys417Asn, Asn440Lys, Val445His, Gly446Ser, Asn450Asp, Leu452Trp, Asn460Lys, Ser477Asn, Thr478Lys, Asn481Lys, Val483del, Glu484Lys, Phe486Pro, Gln498Arg, Asn501Tyr, Tyr505His, Glu554Lys, Ala570Val, Asp614Gly, Pro621Ser, His655Tyr, Ile670Val, Asn679Lys, Pro681Arg,Asn764Lys, Asp796Tyr, Ser939Phe, Gln954His, Asn969Lys, Pro1143Leu) (Arg682Ser, Arg685Ser, Lys986Pro, Val987Pro) (N-terminus) Accession # YP_009724390.1 GCN4-IZ 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SARS-CoV-2 Spike
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.