Learn More
Active Motif Recombinant SARS-CoV-2 NSP1 protein
SDP
Supplier:  Active Motif 81314
Encompass
Recombinant SARS-CoV-2 NSP1 protein was expressed in E.coli cells as the full length protein (accession number YP_009725297.1) with a C-terminal Avi and a C-terminal 6×His tag. The predicted molecular weight of the protein is 19.78 kDa. Recombinant SARS-CoV-2 NSP1 protein is suitable for use in the study of SARS-CoV-2. For more complete and up-to-date information, please visit the Active Motif website.
Catalog No. NC1848737