Learn More
Creative Biomart Recombinant SARS-CoV-2 Spike Protein(S1 Subunit), His-tag, GMP Grade
Supplier:  Creative Biomart SPIKE036THP
Encompass_Preferred
Recombinant SARS-CoV-2 Spike protein GMP Grade(S1 Subunit)(YP_009724390.1)(698 aa) was expressed in HEK293 cells with a C-terminal His Tag.
Catalog No. 50-208-5353