Learn More
Biomatik Corporation Recombinant Severe acute respiratory syndrome coronavirus 2 Spike glycoprotein (S), partial
Supplier:  Biomatik Corporation RPC27085
Encompass_Preferred
Recombinant Severe acute respiratory syndrome coronavirus 2 Spike glycoprotein (S), partial is a purified Recombinant Protein; Coronavirus Protein. Purity: >90% as determined by SDS-PAGE. Host: Yeast. Endotoxin Level: Not Tested. Species: Severe acute respiratory syndrome coronavirus 2 (2019-nCoV) (SARS-CoV-2). Target Name: S. Target Synonyms: E2Peplomer protein. Accession Number: P0DTC2. Expression Region: 16~685aa. Tag Info: N-Terminal 6Xhis-Sumostar-Tagged. Theoretical MW: 88.1kDa. Buffer: Lyophilized from Tris/PBS-based buffer, 6% Trehalose, pH 8.0 Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7235