Learn More
Biomatik Corporation Recombinant Severe acute respiratory syndrome coronavirus 2 Nucleoprotein (N)
Supplier:  Biomatik Corporation RPC27076
Encompass_Preferred
Recombinant Severe acute respiratory syndrome coronavirus 2 Nucleoprotein (N) is a purified Recombinant Protein; Coronavirus Protein. Purity: >85% as determined by SDS-PAGE. Host: Mammalian Cell. Endotoxin Level: Not Tested. Species: Severe acute respiratory syndrome coronavirus 2 (2019-nCoV) (SARS-CoV-2). Target Name: N. Target Synonyms: Nucleocapsid protein. Accession Number: P0DTC9. Expression Region: 1~419aa. Tag Info: Tag-Free. Theoretical MW: 47kDa. Buffer: Lyophilized from 10 mM Tris-HCl, 1 mM EDTA, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7927