Learn More
Biomatik Corporation Recombinant Severe acute respiratory syndrome coronavirus 2 Nucleoprotein (N), Active Protein
Supplier:  Biomatik Corporation RPC27067
Encompass_Preferred
Recombinant Severe acute respiratory syndrome coronavirus 2 Nucleoprotein (N), Active Protein is a purified Active Coronavirus Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Severe acute respiratory syndrome coronavirus 2 (2019-nCoV) (SARS-CoV-2). Target Name: N. Accession Number: P0DTC9. Expression Region: 1~419aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 48.0kDa. Buffer: Lyophilized from a 0.2 um filtered 20 mM Tris-HCl, 0.5 M NaCl, 6% Trehalose, pH 8.0 Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5742