Learn More
Abcam Recombinant Simian Virus 40 SV40 Large T Antigen protein, 5UG
Supplier:  Abcam AB821185UG
Encompass_Preferred
Recombinant Simian Virus 40 SV40 Large T Antigen protein is a Simian Virus 40 Full Length protein, in the 1 to 708 aa range, expressed in Baculovirus, with >95% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE. The predicted molecular weight of ab82118 protein is 83.3 kDa. - Save time and ensure accurate results - use our recombinant SV40 - Antigen protein as a control
Protein/ Peptide/ Enzyme Synonyms: Large T antigen, LT, LT-AG, DNA 3'-5' helicase large T antigen
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1947899