Learn More
Active Motif Recombinant SMARCA2 / BRM (1367-1511) protein
SDP
Supplier:  Active Motif 31849
Encompass
The peptide corresponding to amino acids 1367-1511 that contains the bromodomain sequences of SMARCA2 / BRM (accession number NP_003061.3) was expressed in E. coli and contains an N-terminal His tag and C-terminal FLAG tag with an observed molecular weight of 22.96 kDa. It shows binding specificity for acetylated H3K9, H3K14, H3K9/14, H4K8, H4K12, H4K16 and H4K5/8/12/16.
Catalog No. NC1854361