Learn More
Active Motif RECOMBINANT SMYD2 PRO,1MG
SDP
Supplier:  Active Motif 31901
Encompass
Recombinant SMYD2 protein (accession number NP_064582.2) was expressed in Sf9 cells and contains an N-terminal FLAG-Tag with a molecular weight of 50.7 kDa. The recombinant protein is >90% pure by SDS-PAGE. It is suitable for use in the study of enzyme kinetics, inhibitor screening, and selectivity profiling.
Catalog No. NC1931197