Learn More
Active Motif Recombinant SMYD2 protein
SDP
Supplier:  Active Motif 31497
Encompass
Recombinant SMYD2 protein for use in enzyme kinetics, inhibitor screening, and selectivity profiling.
Catalog No. NC1846110
Edge