Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Zinc metalloproteinase aureolysin (aur)
Supplier:  Biomatik Corporation RPC26039
Encompass_Preferred
Recombinant Staphylococcus aureus Zinc metalloproteinase aureolysin (aur) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Staphylococcus aureus. Target Name: aur. Target Synonyms: Staphylococcus aureus neutral proteinase. Accession Number: P81177. Expression Region: 210~509aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 40.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7931