Learn More
Active Motif Recombinant STAT3 protein
SDP
Supplier:  Active Motif 81095
Encompass
Recombinant human STAT3 protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_644805.1) with an N-terminal FLAG tag. The molecular weight of the protein is 89.2 kDa.
Catalog No. NC1851469