Learn More
Active Motif Recombinant TBP protein
SDP
Supplier:  Active Motif 81114
Encompass
Recombinant human TBP protein was expressed in a baculovirus expression system as the full-length protein (accession number NP_003185.1) with an N-terminal FLAG tag. The molecular weight of the protein is 39 kDa.
Catalog No. NC1851770