Learn More
Active Motif Recombinant Tn5 Transposase, Hyperactive Variant, Efficient DNA Fragmentation, 10 µg
SDP
Catalog No. NC2815712
Encompass
Recombinant Tn5 Transposase is a hyperactive enzyme expressed in *E. coli* designed for efficient tagmentation-based library construction in next-generation sequencing (NGS) applications. Its rapid 5-minute reaction time allows for effective DNA fragmentation and adaptor ligation, resulting in a 9-bp target duplication. With high purity over 90%, it supports various applications such as in vitro transgenic experiments and random library construction. Ideal for researchers seeking reliable and efficient tools in genomic analysis.
Catalog No. NC2815712 Supplier Active Motif Supplier No. 81286
Recombinant Tn5 Transposase is a hyperactive enzyme expressed in *E. coli* designed for efficient tagmentation-based library construction in next-generation sequencing (NGS) applications. Its rapid 5-minute reaction time allows for effective DNA fragmentation and adaptor ligation, resulting in a 9-bp target duplication. With high purity over 90%, it supports various applications such as in vitro transgenic experiments and random library construction. Ideal for researchers seeking reliable and efficient tools in genomic analysis.