Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Tri-ubiquitin/Ub3 K11/K48 branched Protein, CF

Catalog No. UC1148025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UC1148025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. UC1148025 Supplier R&D Systems™ Supplier No. UC1148025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Tri-ubiquitin/Ub3 K11/K48 branched Protein is derived from E. coli. The Recombinant Tri-ubiquitin/Ub3 K11/K48 branched Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P0CG47
For Use With (Application) Bioactivity
Gene ID (Entrez) 7314
Molecular Weight (g/mol) 26 kDa
Quantity 25 μg
Source E. coli-derived human Tri-Ubiquitin protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70 °C as supplied. 12 months, -20 °C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity Tri-Ubiquitin
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer 0.5 mg/mL (20 μm) in 10 mM HEPES pH 7.5
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain. <p>Ub-AQUA analysis:</p><p>K11 + K48: 99%</p><p>All other linkages <= 0.5%</p>
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.