Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Variola CRMB Protein

Catalog No. p-4722859
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
4778CR010 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
4778CR010CF Lyophilized from a 0.2μm filtered solution in PBS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 4778CR010 Supplier R&D Systems™ Supplier No. 4778CR010
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Variola CRMB Protein is derived from NS0. The Recombinant Variola CRMB Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Molecular Weight (g/mol) 36.5 kDa (monomer)
Name Variola CRMB
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived viral CRMB protein Ala23-Leu349,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.